missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol, for analysis, 86-88% wt% aqueous solution, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 295602500

 View more versions of this product

Catalog No. AC295602500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5ppm max
1ppm max
160°C
Glass bottle
1.227
0.005% max.
12 to 14% (K.F.)
Colorless
250mL
56-81-5,7732-18-5
HOCH2CH(OH)CH2OH
01, 502
1,2,3-propanetriol, 1,2,3-trihydroxypropane, glycerin, glycerine, glyceritol, glycerol, glycyl alcohol, osmoglyn, propanetriol, trihydroxypropane
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
86.0 to 88.0%
Glycerol
1.2270g/mL
Authentic
1.4520 to 1.4750
5ppm max
1069 mPa.s (20°C)
290.0°C
18.0°C
86 to 88% wt% Aqueous Solution
C3H8O3
MFCD00004722
15, 4520
Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform and in, fixed and volatile oils
C(C(CO)O)O
92.09
CHEBI:17754
Viscous Liquid
Analysis

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
1,2,3-propanetriol, 1,2,3-trihydroxypropane, glycerin, glycerine, glyceritol, glycerol, glycyl alcohol, osmoglyn, propanetriol, trihydroxypropane
CHEBI:17754
C(C(CO)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 200-289-5

SDS Documents