missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol, 99+%

Manufacturer:  Alfa Aesar A16205AP

 View more versions of this product

Catalog No. AAA16205AP


 

Description

Description

Applications
Glycerol is used as a solvent, sweetener, preservative, antifreezing agent, thickening agent in liqueurs and humectants in the food industry. It is also used in the preparation of pharmaceutical and personal care products, thereby improving the smoothness and provides lubrication. It is an active component in glycerin soap. It finds application in cough syrups, elixirs and expectorants, toothpaste, soaps, mouthwashes, skin care products, shaving cream, hair care products, and water-based personal lubricants. It is used as a starting material for the preparation of nitroglycerin, allyl iodidie, epichlorohydrin and glycerine acetate. In medical field, it is also used as a laxative and a temporary reduction in the internal pressure of the eye. In addition to this, it is involved to aid in casting gradient gels, protein stabilizer and storage buffer component.

Solubility
Miscible with water and alcohol. Partially miscible with ether and ethyl acetate. Immiscible with chloroform.

Notes
Hygroscopic. Incompatible with perchloric acid, lead oxide, acetic anhydride, nitrobenzene, chlorine, peroxides, strong acids and strong bases.
Specifications

Specifications

Glycerol
500mL
≥99%
HOCH2CH(OH)CH2OH
635685
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
182°C (20mmHg)
160°C (320°F)
92.09
56-81-5
C3H8O3
MFCD00004722
14,4484
C(C(CO)O)O
92.094
CHEBI:17754
18°C

Chemical Identifiers

56-81-5
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
CHEBI:17754
C3H8O3
MFCD00004722
753
Safety and Handling

Safety and Handling

GHS H Statement
H319

GHS P Statement
P280-P264-P305+P351+P338-P337+P313

Warning

RTECSNumber : MA8050000

TSCA : TSCA

Documents