Please login to your online account to display your discounted pricing

Glycerol-1-allylether, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Glycerol-1-allylether
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O3
Linear Formula CH2=CHCH2OCH(OH)CH2OH
Formula Weight 132.16
MDL Number MFCD00004721
Synonym 3-(2-Propenyloxy)-1, 2-propanediol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC240671000

  SDS
thermo fisher scientific
AC240671000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC240675000

  SDS
fisher scientific acros
240675000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Viscous Liquid
Formula Weight 132.16
Quantity 100mL
Color Clear colorless to light yellow
Density 1.0700g/mL
Molecular Formula C6H12O3
MDL Number MFCD00004721
CAS 123-34-2
Synonym 3-(2-Propenyloxy)-1, 2-propanediol
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement Causes serious eye irritation. Causes skin irritation.
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Chemical Name or Material Glycerol-1-allylether
Assay Percent Range 98%
Linear Formula CH2=CHCH2OCH(OH)CH2OH
Health Hazard 3 GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes serious eye irritation. Causes skin irritation.