Please login to your online account to display your discounted pricing

Glycerol-1-allylether, 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  240675000

 View more versions of this product

Catalog No. AC240675000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 132.16
Physical Form Viscous Liquid
Quantity 500mL
Color Clear colorless to light yellow
Density 1.0700g/mL
Synonym 3-(2-Propenyloxy)-1, 2-propanediol
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Health Hazard 2 GHS P Statement Causes serious eye irritation. Causes skin irritation.
Health Hazard 3 GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD00004721
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 123-34-2
Chemical Name or Material Glycerol-1-allylether
Linear Formula CH2=CHCH2OCH(OH)CH2OH
Molecular Formula C6H12O3
Assay Percent Range 98%

Specifications

Chemical Name or Material Glycerol-1-allylether
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O3
Linear Formula CH2=CHCH2OCH(OH)CH2OH
Formula Weight 132.16
MDL Number MFCD00004721
Synonym 3-(2-Propenyloxy)-1, 2-propanediol
Accessories

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes serious eye irritation. Causes skin irritation.

SDS & Safety Documents