missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerin, Reagent Grade, LabChem™

Manufacturer:  LabChem LC148501

 View more versions of this product

Catalog No. LC148501



Specifications

Specifications

Glycerin
1.2613g/mL
Passes Test
1.2613g/mL
Colorless
500mL
100
HOCH2CH(OH)CH2OH
Soluble in water
C(C(CO)O)O
92.094
CHEBI:17754
Liquid
Fume; Carbon monoxide; Carbon dioxide
160°C
Poly Bottle
182°C
20°C
56-81-5
C3H8O3
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
Reagent

Chemical Identifiers

56-81-5
92.094
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
C(C(CO)O)O
C3H8O3
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 200-289-5

Recommended Storage : Room Temperature

Documents
Promotions

Promotions