missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerin, Certified, 50% (v/v), LabChem™
$82.83 - $133.67
Chemical Identifiers
| CAS | 56-81-5 |
|---|---|
| Molecular Formula | C3H8O3 |
| Molecular Weight (g/mol) | 92.09 |
| MDL Number | MFCD00004722 |
| InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
| Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
| PubChem CID | 753 |
| ChEBI | CHEBI:17754 |
| IUPAC Name | propane-1,2,3-triol |
| SMILES | OCC(O)CO |
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Specifications
| 56-81-5,7732-18-5 | |
| Carbon monoxide; Carbon dioxide | |
| 1.1g/mL | |
| C3H8O3 | |
| MFCD00004722 | |
| 1.142g/mL ±0.005 | |
| Soluble in water | |
| OCC(O)CO | |
| 92.09 | |
| CHEBI:17754 | |
| 50% (v/v) | |
| Liquid |
| 50,50 | |
| Colorless | |
| Poly Bottle | |
| HOCH2CH(OH)CH2OH | |
| 500 mL | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| 92.09 | |
| Certified | |
| Glycerin |
Safety and Handling
GHS H Statement
Product is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature