missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycerin, 100% Vegetable Based, Multi-Compendial, U.S.P., J.T. Baker™
Supplier: Avantor 214303
Specifications
Glycerin | |
199.00°C | |
C3H8O3 | |
4 L | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
OCC(O)CO | |
92.09 | |
CHEBI:17754 | |
100% Vegetable Based |
56-81-5 | |
Narrow Mouth Amber Glass Bottle | |
MFCD00004722 | |
1L = 1.26kg | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
92.1 | |
Multi-Compendial/USP |
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |