missing translation for 'onlineSavingsMsg'
Learn More

Glybenzcyclamide, 99%

CAS: 10238-21-8 | C23H28ClN3O5S | 494.003 g/mol

Supplier:  Thermo Scientific Chemicals B2145914

 View more versions of this product

Catalog No. AAB2145914


Only null left
Explore available promotions

Description

Description

Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 M).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 μM).

Solubility
Soluble in ethanol (5 mg/mL), DMSO (25 mg/mL), chloroform (1:36), methanol (1:250), and DMF. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications

Specifications

Glybenzcyclamide
171°C to 174°C
MFCD00056625
glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil
ZNNLBTZKUZBEKO-UHFFFAOYSA-N
5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide
3488
494.01
10238-21-8
C23H28ClN3O5S
25 g
Soluble in ethanol (5mg/mL),DMSO (25mg/mL),chloroform (1:36),methanol (1:250),and DMF. Insoluble in water.
COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
494.003
CHEBI:5441
99%

Chemical Identifiers

10238-21-8
494.003
ZNNLBTZKUZBEKO-UHFFFAOYSA-N
3488
5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide
C23H28ClN3O5S
MFCD00056625
glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil
CHEBI:5441
COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-570-6

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only