Please login to your online account to display your discounted pricing

Glutathione Reduced, Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP252125

 View more versions of this product

Catalog No. BP252125

  • / Each
  • / Case of 8 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98%
Melting Point 182°C
Formula Weight 307.3
Physical Form Powder/Solid
Quantity 25g
Merck Index 15, 4511
Molecular Formula C10H17N3O6S
pH 3
Color White
Heavy Metals (as Pb) 10ppm max.
Loss on Drying Report %
Arsenic (As) 1ppm max.
Iron (Fe) 10ppm max.
CAS 70-18-8
ChemAlert Storage Symbol Gray
Recommended Storage 4°C
Assay Percent Range ≥98 %
Specific Rotation -21 to -11° (+ or -)
Ammonium 0.020% max.
Residue after Ignition 0.01% max.
Related Substances 2% max.
Identification Pass Test
Packaging Amber Glass
Sulfate 0.048% max.
Chemical Name or Material Glutathione Reduced
Synonym γ-L-Glutamyl-L-cysteinylglycine

Specifications

Chemical Name or Material Glutathione Reduced
CAS 70-18-8
Physical Form Powder/Solid
Assay ≥98%
Assay Percent Range ≥98 %
Molecular Formula C10H17N3O6S
Formula Weight 307.3
Synonym γ-L-Glutamyl-L-cysteinylglycine
Accessories
 

4°C

SDS & Safety Documents