Please login to your online account to display your discounted pricing

Glutathione Reduced, Fisher BioReagents


Quantity 10g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP252110

 View more versions of this product

Catalog No. BP252110

  • / Each
  • / Case of 8 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98%
Melting Point 182°C
Physical Form Powder/Solid
Formula Weight 307.3
Quantity 10g
Merck Index 15, 4511
Molecular Formula C10H17N3O6S
pH 3
Color White
Arsenic (As) 1ppm max.
Synonym γ-L-Glutamyl-L-cysteinylglycine
Related Substances 2% max.
Specific Rotation -21 to -11° (+ or -)
Ammonium 0.020% max.
Chemical Name or Material Glutathione Reduced
Assay Percent Range ≥98 %
CAS 70-18-8
Heavy Metals (as Pb) 10ppm max.
Loss on Drying Report %
Residue after Ignition 0.01% max.
Sulfate 0.048% max.
ChemAlert Storage Symbol Gray
Identification Pass Test
Iron (Fe) 10ppm max.
Packaging Amber Glass
Recommended Storage 4°C

Specifications

Chemical Name or Material Glutathione Reduced
CAS 70-18-8
Physical Form Powder/Solid
Assay ≥98%
Assay Percent Range ≥98 %
Molecular Formula C10H17N3O6S
Formula Weight 307.3
Synonym γ-L-Glutamyl-L-cysteinylglycine
Accessories
 

4°C

SDS & Safety Documents