missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutathione Reduced, Fisher BioReagents
$389.13 - $1,747.85

Chemical Identifiers
CAS | 70-18-8 |
---|---|
Molecular Formula | C10H17N3O6S |
Molecular Weight (g/mol) | 307.321 |
InChI Key | RWSXRVCMGQZWBV-WDSKDSINSA-N |
Synonym | deltathione, glutathion, glutathione, glutathione-sh, glutinal, isethion, l-glutathione, neuthion, reduced glutathione, tathion |
PubChem CID | 124886 |
ChEBI | CHEBI:16856 |
IUPAC Name | (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid |
SMILES | C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP252110
|
Fisher BioReagents
BP252110 |
10g | Amber Glass |
Each for $389.13
Case of 8 Each for $3,113.04 |
|
BP252125
|
Fisher BioReagents
BP252125 |
25g | Amber Glass |
Each for $621.53
Case of 8 Each for $4,972.24 |
|
BP2521100
|
Fisher BioReagents
BP2521100 |
100g | Amber Glass |
Each for $1,747.85
Case of 4 Each for $6,991.40 |
|
Description
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.Chemical Identifiers
70-18-8 | |
307.321 | |
deltathione, glutathion, glutathione, glutathione-sh, glutinal, isethion, l-glutathione, neuthion, reduced glutathione, tathion | |
CHEBI:16856 | |
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N |
C10H17N3O6S | |
RWSXRVCMGQZWBV-WDSKDSINSA-N | |
124886 | |
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid |
Specifications
70-18-8 | |
White | |
0.01% max. | |
Pass Test | |
C10H17N3O6S | |
15, 4511 | |
10g | |
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N | |
-21 to -11° (+ or -) | |
124886 | |
307.3 | |
≥98% |
3 | |
182°C | |
Report % | |
≥98 % | |
Amber Glass | |
deltathione, glutathion, glutathione, glutathione-sh, glutinal, isethion, l-glutathione, neuthion, reduced glutathione, tathion | |
RWSXRVCMGQZWBV-WDSKDSINSA-N | |
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid | |
307.321 | |
CHEBI:16856 | |
Powder/Solid | |
Glutathione Reduced |
Safety and Handling

Recommended Storage : 4°C