missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutathione Reduced, Fisher BioReagents

Manufacturer:  Fisher BioReagents BP25215

 View more versions of this product

Catalog No. BP25215




Description

Description

Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
Specifications

Specifications

Glutathione Reduced
0.01% max.
Amber Glass
White
5g
70-18-8
C10H17N3O6S
deltathione
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
307.321
CHEBI:16856
Powder/Solid
Pass Test
Report %
-21 to -11° (+ or -)
182°C
3
≥98 %
15, 4511
RWSXRVCMGQZWBV-WDSKDSINSA-N
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
124886
307.3
≥98%

Chemical Identifiers

70-18-8
307.321
deltathione
CHEBI:16856
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
C10H17N3O6S
RWSXRVCMGQZWBV-WDSKDSINSA-N
124886
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
Safety and Handling

Safety and Handling

Recommended Storage : 4°C

SDS Documents
Promotions

Promotions