missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc Glutathione Reduced Crystalline MP Biomedicals

Supplier:  MP Biomedicals Inc 0210181401

 View more versions of this product

Catalog No. ICN10181401



Description

Description

Specifications

Specifications

Glutathione
70-18-8
>98%
MFCD00065939
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
307.321
CHEBI:16856
Powder
Reduced
White
C10H17N3O6S
1 g
RWSXRVCMGQZWBV-WDSKDSINSA-N
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
124886
307.3

Chemical Identifiers

70-18-8
307.321
RWSXRVCMGQZWBV-WDSKDSINSA-N
124886
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
C10H17N3O6S
MFCD00065939
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
CHEBI:16856
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Safety and Handling

Safety and Handling

EINECSNumber : 200-725-4

Recommended Storage : Store at 4°C. Store under nitrogen.

SDS
Documents

Documents

Promotions

Promotions