missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaronitrile 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 157261000

 View more versions of this product

Catalog No. AC157261000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Glutaronitrile
99%
Liquid
99%
C5 H6 N2
94.12
02, 635
15, 4510
Authentic
1.4340 to 1.4360 (20°C, 589nm)
100g
544-13-8
≥98.5%
0.9900g/mL
NC(CH2)3CN
MFCD00001970
04,241
Pentanedinitrile
Glass bottle
Clear colorless to yellow

Chemical Identifiers

544-13-8
C5 H6 N2
94.12
Pentanedinitrile
≥98.5%
99%
NC(CH2)3CN
MFCD00001970
Glutaronitrile, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-861-6

RTECSNumber : YI3500000

TSCA : TSCA