missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glutaronitrile, 99%

Chemical Identifiers

CAS 544-13-8
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00001970
UN Number UN3276
Beilstein 1738385
Merck Index 14,4474
Synonym 1,3-Dicyanopropane; Pentanedinitrile
Chemical Name or Material Glutaronitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2366309 SDS Alfa Aesar™
B2366309
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAB2366318 SDS Alfa Aesar™
B2366318
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
It can be used for the conversion, by exchange, of carboxylic acids to nitriles, by heating in the presence of phosphoric acid. The equilibrium is displaced by cyclization to glutarimide. It is also used in the preparation of dicarbonyl compounds.

Solubility
Soluble in water, alcohol and chloroform.

Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

544-13-8
94.12
UN3276
14,4474
Glutaronitrile
C5H6N2
MFCD00001970
1738385
1,3-Dicyanopropane; Pentanedinitrile

Specifications

10g
C5H6N2
MFCD00001970
1738385
1,3-Dicyanopropane; Pentanedinitrile
−29°C
Glutaronitrile
544-13-8
94.12
UN3276
14,4474
286° to 288°C
112°C (234°F)
99%

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : YI3500000

TSCA : TSCA