Please login to your online account to display your discounted pricing

Glutarimide, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  176250250

 View more versions of this product

Catalog No. AC176250250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Flakes
Assay ≥97.5%
Formula Weight 113.12
Quantity 25g
Color White
Melting Point 154° to 157°C
EINECS Number 214-340-4
RTECS Number MA4000000
Beilstein 21, 382
Name Note 98%
Chemical Name or Material Glutarimide
Assay Percent Range 98%
Molecular Formula C5H7NO2
MDL Number MFCD00006670
CAS 1121-89-7
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Glutarimide
Name Note 98%
CAS 1121-89-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H7NO2
Formula Weight 113.12
MDL Number MFCD00006670