Please login to your online account to display your discounted pricing

Glutarimide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Glutarimide
Name Note 98%
CAS 1121-89-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H7NO2
Formula Weight 113.12
MDL Number MFCD00006670
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176250250

  SDS
fisher scientific acros
176250250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Flakes
Assay ≥97.5%
Formula Weight 113.12
Quantity 25g
Color White
Melting Point 154° to 157°C
EINECS Number 214-340-4
RTECS Number MA4000000
Beilstein 21, 382
Name Note 98%
Chemical Name or Material Glutarimide
Assay Percent Range 98%
Molecular Formula C5H7NO2
MDL Number MFCD00006670
CAS 1121-89-7
Infrared Spectrum Authentic
Packaging Glass bottle