missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC233281000
Specifications
Glutaric dialdehyde, electron microscopy grade | |
Glass bottle | |
1.06 | |
Undesignated | |
100mL | |
Purified | |
C5H8O2 | |
MFCD00007025 | |
01,411 | |
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
25 to 27 wt% |
1.0600g/mL | |
1.373 | |
101°C | |
-5.0°C | |
3.1 to 4.5 (25°C) | |
111-30-8,7732-18-5 | |
OHC(CH2)3CHO | |
01, 776 | |
15, 4508 | |
Solubility in water: soluble | |
C(CC=O)CC=O | |
100.12 | |
CHEBI:64276 | |
Solution | |
E.M. |
Chemical Identifiers
111-30-8 | |
100.12 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |
C5H8O2 | |
MFCD00007025 | |
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide | |
CHEBI:64276 | |
C(CC=O)CC=O |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Very t
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 203-856-5