missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaric Dialdehyde, 50% in Water, Spectrum™
$228.40 - $5,478.71
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610722
|
Spectrum Chemical Mfg Cor
G3060100ML |
100 mL | Amber Glass Bottle |
Each for $228.40
|
|
18610723
|
Spectrum Chemical Mfg Cor
G3060500ML |
500 mL | Amber Glass Bottle |
Each for $472.31
|
|
18610724
|
Spectrum Chemical Mfg Cor
G30604LT |
4 L | Amber Glass Bottle |
Each for $1,201.54
|
|
18610725
|
Spectrum Chemical Mfg Cor
G306020LTBL |
20 L | Poly Drum |
Each for $5,478.71
|
|
Chemical Identifiers
111-30-8 |
Specifications
Amber Glass Bottle | |
100 mL | |
111-30-8,7732-18-5 | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.