missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaric Dialdehyde, 50% in Water, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor G306020LTBL
Specifications
111-30-8,7732-18-5 | |
Poly Drum | |
20 L | |
Reagent |
Chemical Identifiers
111-30-8 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.