missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric anhydride, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1115222

 View more versions of this product

Catalog No. AAA1115222



Description

Description

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
Specifications

Specifications

Glutaric anhydride
108-55-4
98%
MFCD00006679
Dihydro-2H-pyran-2,6(3H)-dione; Pentanedioic anhydride
C1CC(=O)OC(=O)C1
114.1
114.1
52°C to 57°C
100g
98%
C5H6O3
110051
VANNPISTIUFMLH-UHFFFAOYSA-N
oxane-2,6-dione
7940
291°C
178°C (352°F)

Chemical Identifiers

108-55-4
114.1
VANNPISTIUFMLH-UHFFFAOYSA-N
7940
C1CC(=O)OC(=O)C1
C5H6O3
MFCD00006679
Dihydro-2H-pyran-2,6(3H)-dione; Pentanedioic anhydride
oxane-2,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
H318-H312-H335-H315
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : MA3850000

TSCA : TSCA

SDS Documents