Learn More
Glutaric anhydride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119971000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric anhydride | |
108-55-4 | |
White | |
173°C | |
94% min. (GC) | |
C5H6O3 | |
100 g | |
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech | |
VANNPISTIUFMLH-UHFFFAOYSA-N | |
oxane-2,6-dione | |
7940 | |
95% |
97% | |
46.0°C to 57.0°C | |
290.0°C | |
Authentic | |
Glass bottle | |
MFCD00006679 | |
17, V,11, 9 | |
Solubility in water: hydrolysis. Other solubilities: soluble in benzene, toluene, acetone and, chloroform | |
C1CC(=O)OC(=O)C1 | |
114.1 | |
114.1 | |
Crystalline Powder or Crystals |
Chemical Identifiers
108-55-4 | |
114.1 | |
VANNPISTIUFMLH-UHFFFAOYSA-N | |
7940 | |
C1CC(=O)OC(=O)C1 |
C5H6O3 | |
MFCD00006679 | |
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech | |
oxane-2,6-dione |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 203-593-6
RTECSNumber : MA3850000
TSCA : TSCA
RUO – Research Use Only