Please login to your online account to display your discounted pricing

Glutaric anhydride, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Glutaric anhydride
CAS 108-55-4
Assay ≥88% (morpholine coupling)
Assay Percent Range 90%
Molecular Formula C5H6O3
Formula Weight 114.1
MDL Number MFCD00006679
Synonym Tetrahydropyran-2, 6-dione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388040500

  SDS
thermo fisher scientific
AC388040500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388042500

  SDS
thermo fisher scientific
AC388042500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88% (morpholine coupling)
Formula Weight 114.1
Physical Form Crystalline Powder
Quantity 50g
Color Beige to light brown
Melting Point 46° to 57°C
Synonym Tetrahydropyran-2, 6-dione
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H6O3
Assay Percent Range 90%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
MDL Number MFCD00006679
Packaging Glass bottle
CAS 108-55-4
Chemical Name or Material Glutaric anhydride
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.