missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119960250

 View more versions of this product

Catalog No. AC119960250



Description

Description

Specifications

Specifications

Glutaric acid
Glass Bottle
Orange
25g
110-94-1
C5H8O4
MFCD00004410
01,412
1,3-propanedicarboxylic acid, 1,5-pentanedioic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di, glutarate, glutaric acid, n-pyrotartaric acid, pentandioic acid, propane-1,3-dicarboxylic acid, unii-h849f7n00b
JFCQEDHGNNZCLN-UHFFFAOYSA-N
pentanedioic acid
743
132.12
99%
Authentic
302.0°C
92.0°C to 99.0°C
99%
99%
HO2C(CH2)3CO2H
02, 631
15, 4509
Solubility in water: 430g/L (20°C). Other solubilities: freely soluble in abs. alcohol and ether, soluble in benzene and chloroform
OC(=O)CCCC(O)=O
132.12
CHEBI:17859
Crystalline Powder

Chemical Identifiers

110-94-1
132.12
JFCQEDHGNNZCLN-UHFFFAOYSA-N
743
pentanedioic acid
C5H8O4
MFCD00004410
1,3-propanedicarboxylic acid, 1,5-pentanedioic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di, glutarate, glutaric acid, n-pyrotartaric acid, pentandioic acid, propane-1,3-dicarboxylic acid, unii-h849f7n00b
CHEBI:17859
OC(=O)CCCC(O)=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISO

GHS Signal Word: Warning

EINECSNumber : 203-817-2

RTECSNumber : MA3740000

TSCA : TSCA

SDS Documents
Product Certifications