Learn More
Glutaric acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119965000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric acid | |
110-94-1 | |
Orange | |
Authentic | |
Plastic Bottle | |
HO2C(CH2)3CO2H | |
500 g | |
01,412 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
pentanedioic acid | |
743 | |
132.12 | |
Crystalline Powder |
99% | |
92.0°C to 99.0°C | |
302.0°C | |
99% | |
C5H8O4 | |
MFCD00004410 | |
02, 631 | |
15, 4509 | |
Solubility in water: 430g/L (20°C). Other solubilities: freely soluble in abs. alcohol and ether, soluble in benzene and chloroform | |
OC(=O)CCCC(O)=O | |
132.12 | |
CHEBI:17859 | |
99% |
Chemical Identifiers
110-94-1 | |
132.12 | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
743 | |
pentanedioic acid |
C5H8O4 | |
MFCD00004410 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di | |
CHEBI:17859 | |
OC(=O)CCCC(O)=O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 203-817-2
RTECSNumber : MA3740000
TSCA : TSCA
RUO – Research Use Only