missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutardialdehyde Solution 25%, For Electron Microscopy, MilliporeSigma™

Manufacturer:  MilliporeSigma 1042392500

 View more versions of this product

Catalog No. M1042392500



Description

Description

For electron microscopy
Specifications

Specifications

Glutardialdehyde solution
Colorless liquid
25%
C5H8O2
UN 2922
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
0.27 hPa at 20°C
100.117
CHEBI:64276
2.5L
111-30-8
Characteristic odor
Glass bottle
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
C(CC=O)CC=O
pentanedial
3485
Liquid

Chemical Identifiers

111-30-8
100.117
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
C(CC=O)CC=O
C5H8O2
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
Safety and Handling

Safety and Handling

H302 + H332: Harmful if swallowed or if inhaled
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
H400: Very toxic to aquatic life.

DOTInformation : DOT Class: 8 (6.1), Packaging Group: II

Documents
Promotions

Promotions