missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutardialdehyde Solution 25%, For Electron Microscopy, acc. to P.J. Anderson, EMD Millipore™

Manufacturer:  Emd Millipore Corporation 1121790025

 View more versions of this product

Catalog No. M1121790025


 

Specifications Description & Specifications

Specifications

Glutardialdehyde Solution
Colorless liquid
Characteristic odor
25%
UN 2922
Purified and filled under nitrogen
111-30-8
Liquid
Glass bottle
0.27 hPa at 20°C

Chemical Identifiers

111-30-8
UN 2922
Glutardialdehyde Solution, Purified and filled under nitrogen

Safety and Handling

H302 + H332: Harmful if swallowed or if inhaled
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
H400: Very toxic to aquatic life.

DOTInformation : DOT Class: 8 (6.1), Packaging Group: II