Please login to your online account to display your discounted pricing

Alfa Aesar™ Glutaranilic acid, 97%

$295.54 - $295.54

Chemical Identifiers

Chemical Name or Material Glutaranilic acid
CAS 5414-99-3
Molecular Formula C11H13NO3
Formula Weight 207.23
MDL Number MFCD00021788
Beilstein 2807261
Synonym Pentanedioic acid mono(N-phenyl)amide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0895514

 
alfa aesar™
L0895514
25g Each for $295.54
Description & Specifications

Specifications

Formula Weight 207.23
Chemical Name or Material Glutaranilic acid
Melting Point 124° to 128°C
Assay 97%
Molecular Formula C11H13NO3
MDL Number MFCD00021788
CAS 5414-99-3
Synonym Pentanedioic acid mono(N-phenyl)amide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Beilstein 2807261
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.