missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaraldehyde Solution, 25% (Certified), Fisher Chemical

Manufacturer:  Thermo Scientific O29571

 View more versions of this product

Catalog No. O2957-1




Specifications

Specifications

Glutaraldehyde Solution, 25%
Vapor Pressure: 16.4mmHg at 20°C
1.060g/cm³
24.0 to 26.5 %
HC(O)(CH2)3CHO
MFCD00007025
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
16.4mmHg at 20°C
100.117
CHEBI:64276
Liquid
Certified
-5°C
1L
111-30-8,7732-18-5
Pass Test
C5H8O2
Glass Bottle
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
C(CC=O)CC=O
pentanedial
3485
100.12
24.0 to 26.5%
100°C
Undesignated

Chemical Identifiers

111-30-8
100.117
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
C(CC=O)CC=O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1

SIGNAL WORD: DANGER!

DOTInformation : DOT Class 6.1, : Poison

SDS Documents