Please login to your online account to display your discounted pricing

Glutaraldehyde Solution (25%/Certified/BioReagent), Fisher BioReagents


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BP25484

 View more versions of this product

Catalog No. BP25484

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 100.12
Boiling Point 100°C
Melting Point −5°C
Recommended Storage KEEP REFRIGERATED or in a very cool place to retard polymerisation
Quantity 4L
Merck Index 15, 4508
DOT Information DOT Class 6.1, : Poison
Density 1.060g/cm³
Physical Form Liquid
Assay 24.0 to 26.5%
Vapor Pressure 16.4mmHg at 20°C
Viscosity [NULL]
Color Clear
Additional Information Vapor Pressure: 16.4mmHg at 20°C
Name Note 25%
Assay Percent Range 24.0 to 26.5 %
Packaging Glass Bottle
Linear Formula HC(O)(CH2)3CHO
CAS 111-30-8
Health Hazard 1 SIGNAL WORD: DANGER!
Health Hazard 2 DANGER!
CAS 7732-18-5
Health Hazard 3 Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1
Chemical Name or Material Glutaraldehyde Solution
Identification Pass Test
Molecular Formula C5H8O2
ChemAlert Storage Symbol Blue
Synonym Glutaral

Specifications

Chemical Name or Material Glutaraldehyde Solution
Name Note 25%
CAS 111-30-8
CAS 7732-18-5
Purity Grade Certified Grade
Assay 24.0 to 26.5%
Assay Percent Range 24.0 to 26.5 %
Molecular Formula C5H8O2
Linear Formula HC(O)(CH2)3CHO
Formula Weight 100.12
Synonym Glutaral
Recommended Storage KEEP REFRIGERATED or in a very cool place to retard polymerisation

KEEP REFRIGERATED or in a very cool place to retard polymerisation

Accessories
 

SIGNAL WORD: DANGER!

Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1

DANGER!

KEEP REFRIGERATED or in a very cool place to retard polymerisation