missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaraldehyde Solution (25%/Certified/BioReagent), Fisher BioReagents

$896.38 - $896.38

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym 1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP25484
SDS View Documents Promotion Details
Fisher BioReagents
BP25484
4L Glass Bottle
Each for $896.38
Case of 4 Each for $3,585.52
 
Description

Description

Specifications

Chemical Identifiers

111-30-8
100.12
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide
CHEBI:64276
O=CCCCC=O

Specifications

4L
111-30-8,7732-18-5
Pass Test
C5H8O2
HC(O)(CH2)3CHO
15, 4508
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
O=CCCCC=O
100.12
CHEBI:64276
Liquid
Certified
-5°C
Glutaraldehyde Solution, 25%
Vapor Pressure: 16.4mmHg at 20°C
1.060g/cm³
24.0 to 26.5 %
Glass Bottle
MFCD00007025
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide
16.4mmHg at 20°C
pentanedial
3485
100.12
24.0 to 26.5%
100°C
Undesignated
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1

SIGNAL WORD: DANGER!

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : KEEP REFRIGERATED or in a very cool place to retard polymerisation

SDS
Product Certifications