missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaraldehyde, 3%, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR32818001500A

 View more versions of this product

Catalog No. R3281800150



Specifications

Specifications

Glutaraldehyde
85.0,11.6,0.84,0.52
dihydrogen monoxide, dihydrogen oxide
Poly natural
Colorless
7732-18-5,111-30-8,7558-79-4,10049-21-5
89.0,12.07,0.87,0.54
Liquid
3%
1.50L

Chemical Identifiers

7732-18-5
dihydrogen monoxide, dihydrogen oxide
Documents