Please login to your online account to display your discounted pricing

Glutaraldehyde, 3%, Ricca Chemical

$20.03 - $20.03

Chemical Identifiers

Chemical Name or Material Glutaraldehyde
CAS 7732-18-5
CAS Min % 85.0
CAS Max % 89.0
CAS 111-30-8
CAS Min % 11.6
CAS Max % 12.07
CAS 7558-79-4
CAS Min % 0.84
CAS Max % 0.87
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R3281800360

 
ricca chemical company
RIR3281800360A
Poly natural 360mL Each for $20.03

R3281800150

 
ricca chemical company
RIR32818001500A
Poly natural 1.50L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 360mL
Concentration or Composition Notes 3%
Packaging Poly natural
CAS Max % 89.0
CAS 111-30-8
CAS 7558-79-4
CAS Min % 0.84
CAS Max % 0.54
CAS Min % 85.0
CAS Max % 12.07
CAS Min % 11.6
CAS 10049-21-5
CAS Min % 0.52
CAS Max % 0.87
Chemical Name or Material Glutaraldehyde
CAS 7732-18-5