Please login to your online account to display your discounted pricing

Glutaraldehyde, 25% in H2O, Practical, J.T.Baker™

$135.07 - $1,044.47

Chemical Identifiers

Chemical Name or Material Glutaraldehyde
CAS 111-30-8
Purity Grade Practical Grade
Molecular Formula OHC(CH2)3CHO
Formula Weight 100.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002921

 
avantor performance
M75209
Narrow Mouth Amber Glass Bottle 4L Case of 4 Each for $1,044.47

02002920

 
avantor performance
M75207
Narrow Mouth Amber Glass Bottle 500mL Each for $135.07
Description & Specifications

Specifications

Purity Grade Practical Grade
Formula Weight 100.12
Chemical Name or Material Glutaraldehyde
Molecular Formula OHC(CH2)3CHO
CAS 111-30-8
Assay 25% in H2O
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.09kg
Quantity 4L