missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaraldehyde, 25% aqueous solution, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219519980
Specifications
Glutaraldehyde | |
Colorless | |
111-30-8 | |
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide | |
C(CC=O)CC=O | |
100.117 | |
CHEBI:64276 | |
Liquid |
1.061g/mL | |
100mL | |
C5H8O2 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.11 |
Chemical Identifiers
111-30-8 | |
100.117 | |
1,5-pentanedial, aldesan, cidex, glutaral, glutaraldehyde, glutardialdehyde, glutaric aldehyde, glutaric dialdehyde, glutarol, sonacide | |
CHEBI:64276 | |
C(CC=O)CC=O |
C5H8O2 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |