Please login to your online account to display your discounted pricing

Glutaraldehyde, 25% Aqueous Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$354.42 - $2,631.27

Chemical Identifiers

Chemical Name or Material Glutaraldehyde
CAS 111-30-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula OHC(CH2)3CHO
Formula Weight 100.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002918

 
avantor performance
212707
Poly Pail 19L Each for $2,631.27

02002919

 
avantor performance
212703
Narrow Mouth Amber Glass Bottle 4L Each for $932.21

02002922

 
avantor performance
212701
Narrow Mouth Amber Glass Bottle 500mL Each for $354.42
Description & Specifications

Specifications

Chemical Name or Material Glutaraldehyde
Quantity 19L
CAS 111-30-8
Purity Grade BAKER ANALYZED™ Reagent
Assay 25% Aqueous Solution
Molecular Formula OHC(CH2)3CHO
Formula Weight 100.12
Packaging Poly Pail
Specific Gravity 1L = 1.09kg