Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Glutamyl Transferase, γ Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, flow cytometry and immunoprecipitation experiments.

$308.98 - $1,473.14

Specifications

Antigen Glutamyl Transferase, gamma Ab-1
Clone 138H11
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1272P

 
thermo scientific™ lab vision™
MS1272P
200μg/mL 1mL Each for $1,421.29

MS1272P0

 
thermo scientific™ lab vision™
MS1272P0
200μg/mL 100µL Each for $308.98

MS1272P1

 
thermo scientific™ lab vision™
MS1272P1
200μg/mL 500µL Each for $869.43

MS1272P1ABX

 
thermo scientific™ lab vision™
MS1272P1ABX
1mg/mL 100µL Each for $998.82

MS1272PABX

 
thermo scientific™ lab vision™
MS1272PABX
1mg/mL 200µL Each for $1,473.14
Description & Specifications

Specifications

Antigen Glutamyl Transferase, gamma Ab-1
Clone 138H11
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Immunogen Purified human kidney GGT
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry, western blotting, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Glutamyl Transferase, γ Ab-1, Mouse Monoclonal Antibody.

Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the γ-glutamly moiety of γ-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.

Host Species: Mouse

Clone: 138H11

Isotype: IgG1

Species Reactivity: Human. Does not react with pig and cow. Others not known.

Epitope: Not determined

Immunogen: Purified human kidney GGT

Molecular Weight: 90 kDa

Positive Control: CAKi renal tumor cells, Renal cell CA or kidney

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Flow Cytometry
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO