Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Glutamyl Transferase, γ Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, flow cytometry and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1272PABX

 View more versions of this product

Catalog No. MS1272PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Glutamyl Transferase, gamma Ab-1
Regulatory Status RUO
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human kidney GGT
Isotype IgG1
Quantity 200µL
Format Concentrated
Host Species Mouse
Clone 138H11
Research Discipline Enzyme
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal

Ensure accurate, reproducible results in immunohistochemistry, western blotting, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Glutamyl Transferase, γ Ab-1, Mouse Monoclonal Antibody.

Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the γ-glutamly moiety of γ-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.

Host Species: Mouse

Clone: 138H11

Isotype: IgG1

Species Reactivity: Human. Does not react with pig and cow. Others not known.

Epitope: Not determined

Immunogen: Purified human kidney GGT

Molecular Weight: 90 kDa

Positive Control: CAKi renal tumor cells, Renal cell CA or kidney

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Flow Cytometry
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO