missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1839006

 View more versions of this product

Catalog No. AAA1839006



Description

Description

Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

Solubility
Soluble in methanol. Insoluble in water..

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications

Specifications

Glutaconic aldehyde dianilide hydrochloride
1497-49-0
C17H17ClN2
5 g
Hygroscopic
Soluble in methanol. Insoluble in water..
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1
284.79
284.79
mixture of isomers
∼124°C to 132°C
MFCD00012630
3916924
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
VUCMMJBDNXZQDJ-ZUJIUJENSA-N
N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
5702667
98%

Chemical Identifiers

1497-49-0
284.79
VUCMMJBDNXZQDJ-ZUJIUJENSA-N
5702667
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1
C17H17ClN2
MFCD00012630
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 216-094-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only