Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1, Rat Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  RT102P1ABX

 View more versions of this product

Catalog No. RT102P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Applications Immunoprecipitation
Applications Western Blotting
Antigen Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1
Regulatory Status RUO
Format Concentrated
Quantity 100µL
Isotype IgG2a
Immunogen Purified glucose regulated protein 94 (grp94) from chicken oviducts
Host Species Rat
Clone 9G10.F8.2
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Steroids Hormones and Growth Factors
Concentration 1mg/mL
Type Primary
Classification Monoclonal
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1, Rat Monoclonal Antibody.

Grp's (Glucose-Regulated Proteins) are a class of proteins synthesized in cells in response to glucose starvation. Glucose-regulated protein 94 (grp94) or tumor rejection antigen (gp96) shows a high degree of sequence homology with the heat shock protein 90 (hsp90). Grp94 has been briefly studied as a prognostic factor in breast cancer.

Host Species: Rat

Clone: 9G10.F8.2

Isotype: IgG2a

Species Reactivity: Human, Monkey, Horse, Cow, Sheep, Pig, Dog, Rabbit, Rat, Mouse, Hamster, Guinea Pig, Chicken and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Purified glucose-regulated protein 94 (grp94) from chicken oviducts

Molecular Weight: 94 kDa

Positive Control: SKBR-3 cells, Tonsil and Breast Carcinomas.

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)

Note:

USA: RUO; Int'l: RUO