Please login to your online account to display your discounted pricing
Thermo Scientific™ Glucose Hexokinase Reagents
Determine quantity of glucose in human serum, plasma or urine in vitro with Thermo Scientific™ Glucose Hexokinase Reagents.
$198.97 - $198.97
Specifications
CE Marker Yes
Methods Compatibility Endpoint
Storage Requirements 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    
TR15421
thermo scientific™
TR15421
Infinity Glucose Hexokinase 2 x 125mL Pack for $198.97
Description & Specifications
Specifications
CE Marker Yes
Methods Compatibility Endpoint
Storage Requirements 2° to 8°C
Determine quantity of glucose in human serum, plasma or urine in vitro with Thermo Scientific™ Glucose Hexokinase Reagents.
Diabetes mellitus is a group of metabolic disorders of carbohydrate metabolism in which glucose is underutilized, producing hyperglycemia. Measurement of blood glucose levels plays a key role in the diagnosis and management of the disease. Increased glucose levels may also be observed with hyperactivity of the thyroid, pituitary, or adrenal glands. Decreased levels are present in cases of insulin overdose, insulin secreting tumors, myxedema, hypopituitarism, hypoadrenalism, and conditions interfering with glucose absorption.
  • Up to 18 months of stability from the date of manufacture