missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gluconic acid, potassium salt, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 229320010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Gluconic acid, potassium salt | |
299-27-4 | |
Authentic | |
Plastic bottle | |
HOCH2[CH(OH)]4CO2K | |
1kg | |
+14.10° (21°C c=1.2,H2O) | |
potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal | |
HLCFGWHYROZGBI-JJKGCWMISA-M | |
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate | |
16760467 | |
99% |
99% | |
3% max. (105°C, 4 hrs) (vacuum) | |
98.5% min. (C6H11KO7) On dried substance | |
C6H11KO7 | |
MFCD00064211 | |
15, 4492 | |
+ 14.1 | |
Solubility in water: soluble in water. Other solubilities: practically insoluble in abs. alcohol,ether,,benzene and chloroform | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+] | |
234.25 | |
234.25 |
Safety and Handling
EINECSNumber : 206-074-2
RTECSNumber : LZ5230000
TSCA : TSCA
RUO â Research Use Only