missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gluconic Acid, 50 wt. % Solution in Water, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 119890025

 View more versions of this product

Catalog No. AC119890025



Description

Description

Specifications

Specifications

Gluconic acid
C6H12O7
1000ppm max.
15, 4492
1.23g/mL
C(C(C(C(C(C(=O)O)O)O)O)O)O
196.155
CHEBI:33198
Crystalline Powder or Crystals
10ppm max.
3% max.
1.22 to 1.25 (20°C)
White to Yellow
526-95-4,7732-18-5
MFCD00004240
03, 542
d-gluconate, d-gluconic acid, dextronic acid, glosanto, gluconate, gluconic acid, gluconic acid, d, glycogenic acid, maltonic acid, pentahydroxycaproic acid
RGHNJXZEOKUKBD-SQOUGZDYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
10690
196.16
49 to 55% (Titrimetry other)
Glass bottle
3 % max.
1000ppm max.
2.5L

Chemical Identifiers

526-95-4
196.155
RGHNJXZEOKUKBD-SQOUGZDYSA-N
10690
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
C6H12O7
MFCD00004240
d-gluconate, d-gluconic acid, dextronic acid, glosanto, gluconate, gluconic acid, gluconic acid, d, glycogenic acid, maltonic acid, pentahydroxycaproic acid
CHEBI:33198
C(C(C(C(C(C(=O)O)O)O)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 208-401-4

RTECSNumber : LZ5057100

TSCA : TSCA

SDS Documents