missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glucagon Mouse, eFluor 570, Clone: ICACLS, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $404.55

Specifications

Antigen Glucagon
Clone ICACLS
Host Species Murine
Gene Alias GCG
Species Reactivity Human, Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123645 Life Technologies
LS41974380
25μg Each for $160.95
501123646 Life Technologies
LS41974382
100μg Each for $404.55
Specifications

Description

This ICACLS monoclonal antibody reacts with human, mouse, and rat glucagon. Glucagon is a peptide hormone secreted by the pancreatic alpha cells found within the islets of Langerhans. Glucagon functions as an antagonist to insulin and homeostatic regulation of blood glucose levels are maintained by the critical balance between these two hormones. Glucagon acts in the liver to convert glycogen to glucose by promoting gluconeogenesis and glycogenolysis. Secretion of glucagon is stimulated by hypoglycemia, epinephrine, acetylcholine, and cholecystokinin, while secretion is inhibited by insulin and somatostatin. Abnormally high levels of glucagon are found in pancreatic tumors.
Description & Specifications

Specifications

Glucagon
Murine
Human, Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ICACLS
GCG
Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin)
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL