Please login to your online account to display your discounted pricing

Glucagon Mouse, Biotin, Clone: ICACLS, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $397.30

Specifications

Antigen Glucagon
Clone ICACLS
Host Species Murine
Gene Alias GCG
Species Reactivity Human, Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122592

 
life technologies
LS13974380
25μg Each for $146.45

501122593

 
life technologies
LS13974382
100μg Each for $397.30
Description & Specifications

Specifications

Antigen Glucagon
Clone ICACLS
Host Species Murine
Gene Alias GCG
Species Reactivity Human, Mouse, Rat
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This ICACLS monoclonal antibody reacts with human, mouse, and rat glucagon. Glucagon is a peptide hormone secreted by the pancreatic alpha cells found within the islets of Langerhans. Glucagon functions as an antagonist to insulin and homeostatic regulation of blood glucose levels are maintained by the critical balance between these two hormones. Glucagon acts in the liver to convert glycogen to glucose by promoting gluconeogenesis and glycogenolysis. Secretion of glucagon is stimulated by hypoglycemia, epinephrine, acetylcholine, and cholecystokinin, while secretion is inhibited by insulin and somatostatin. Abnormally high levels of glucagon are found in pancreatic tumors.