Please login to your online account to display your discounted pricing

Alfa Aesar™ Glucagon-Like Peptide-2, Human


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66884MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66884MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Sequence GLP-2, human
Molecular Weight 3922.40
Quantity 1mg
Cross Reactivity Human
Storage Requirements -30° to -10°C
Product Type Glucagon-Like Peptide-2, human
Format Solid
Regulatory Status RUO
Purification Method HPLC
Common Name Preproglucagon (126-158), human