Please login to your online account to display your discounted pricing

Alfa Aesar™ Glucagon-Like Peptide-1, Human

For cellular and molecular biology applications

Specifications

Product Type Glucagon-Like Peptide-1, human
Cross Reactivity Human
Molecular Weight 4169.60
Storage Requirements -30° to -10°C
Sequence Preproglucagon (72-108)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66108EXD

 
alfa aesar™
J66108EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66108MCR

 
alfa aesar™
J66108MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Glucagon-Like Peptide-1, human
Cross Reactivity Human
Molecular Weight 4169.60
Storage Requirements -30° to -10°C
Sequence Preproglucagon (72-108)
Format Solid
Common Name GLP-1
Regulatory Status RUO
Purification Method HPLC