Please login to your online account to display your discounted pricing

Alfa Aesar™ Glu-Ile-Leu-Glu-Val-Pro-Ser-Thr

For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66722-MCR

 View all options for this product

Catalog No. AAJ66722MCR

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Molecular Weight 886.98
Purification Method HPLC
Quantity 1mg
Format Solid
Sequence E-I-L-E-V-P-S-T
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Glu-Ile-Leu-Glu-Val-Pro-Ser-Thr