Please login to your online account to display your discounted pricing

Alfa Aesar™ [Glp&superscript_4;] Myelin Basic Protein (4-14)

For cellular and molecular biology applications

Specifications

Product Type [Glp4] Myelin Basic Protein (4-14)
Molecular Weight 1373.60
Storage Requirements -30° to -10°C
Sequence Glp-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66467EXD

 
alfa aesar™
J66467EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66467MCR

 
alfa aesar™
J66467MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Glp4] Myelin Basic Protein (4-14)
Molecular Weight 1373.60
Storage Requirements -30° to -10°C
Sequence Glp-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
Format Solid
Regulatory Status RUO
Purification Method HPLC