missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ GloWear™ 8205HL Type R Class 2 Super Econo Mesh Vest
Specifications
Specifications
| Certifications/Compliance | ANSI-compliant polyester mesh |
| Color | Lime |
| Size | X-Small |
| Material | Polyester |
| For Use With (Application) | Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Baggage Handlers, Emergency Response, Warehousing/Distribution |